Order Entry
Belgium
ContactUsLinkComponent
 
 
 
 
Paraformaldehyde 8% (w/v) in aqueous solution methanol-free
 
 
 
 
Paraformaldehyde 8% (w/v) in aqueous solution methanol-free

 

  • Appearance (Color):
    Clear colorless
  • Form:
    Liquid
  • Ash:
    ≤0.01% (Paraformaldehyde content)
  • Comment:
    Manufactured from 95% Paraformaldehyde prills
  • Concentration:
    7.5 to 8% (Formaldehyde)
  • Trace Metal:
    Iron (Fe) ≤2.0% (Paraformaldehyde content)

 

Frequently Bought Together