ContactUsLinkComponent
Multi-element ICH Q3D and USP 232 kit for metals analysis, Specpure®
Multi-element ICH Q3D and USP 232 kit for metals analysis, Specpure®
Catalog # 47384.6C
CAS Number:  
Multi-element ICH Q3D and USP 232 kit for metals analysis, Specpure®
Catalog # 47384.6C
CAS Number:  
Specifications
  • Pk:
    5
  • Description:
    Multi-Element ICH Q3D & USP 232 Kit for Metals Analysis
  • Environmentally Preferable:
  • Pk Info:
    5× 100 ml
Specifications
About this item
This kit includes the following five items:
47318-AE, Multi-Element ICH/USP Oral Standard Solution A
47370-AE, Multi-Element ICH/USP Oral Standard Solution B
47387-AE, Multi-Element ICH/USP Oral Standard Solution C
47325-AE, Multi-Element ICH/USP Oral Standard Solution D
47358-AE, Multi-Element Pharma Internal Standard Solution 1