ContactUsLinkComponent
Disposable particle filtering respirators, FFP3, P309
Disposable particle filtering respirators, FFP3, P309
  PRTWP309WHR
 :  Portwest
undefined
Disposable particle filtering respirators, FFP3, P309
  PRTWP309WHR
 :  Portwest
  • Type:
    P309
  • Pk:
    2
  • With valve:
    +
  • Class:
    FFP3
This mask with valve protects against a high concentration of dust, water based mist and fumes.
  • High performance exhalation valve to reduce heat and provide comfort when used in hot and humid conditions
  • Large inner space to help the user to breathe more easily
  • Adjustable straps for a secure fit
The P301 can be used in concentrations of contaminants up to 20 times the WEL.
 : EN 149:2001 + A1:2009.
 : Blister pack: Two mask per pack.