Dichloromethane, anhydrous stabilised for applications requiring low water content <0.0010% H₂O, Riedel-de Haën™
Catalog # HONC66204-100ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Bottle, septum-sealed
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (GC):Min. 99.9 %
- Water (Karl Fischer):Max. 0.0010 %
- Non-volatile matter:Max. 0.0001 %
Specification Test Results
About this item
Synonyms: Methylene chloride
:
Boiling Pt:39…41 °C (1.013 hPa)
Melting Pt:–97 °C
Density:1.32…1.33 g/cm3 (20 °C)
EINECS:200-838-9
ADR:6.1,III
:Packed in septum-sealed bottle.
:Stabilised with amylene
:84.93 g/mol
Storage Temperature:Ambient
UN:1593
:GHS07
:GHS08
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881