Order Entry
Belgium
ContactUsLinkComponent
n-Heptane for gas chromatography, Burdick & Jackson™
n-Heptane for gas chromatography, Burdick & Jackson™
Catalog # HONC65836-7L-RC
CAS Number:  
undefined
n-Heptane for gas chromatography, Burdick & Jackson™
Catalog # HONC65836-7L-RC
CAS Number:  

Specifications

  • Pack type:
    Stainless steel can
  • Pk:
    7 L
  • Seq:
    5202

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Suitability for residue analysis:
    Max. 5 ng/l

Specification Test Results

About this item

 :
 :100.20 g/mol
Boiling Pt:98.2…98.4 °C (1.000 hPa)
Melting Pt:–90.5 °C
Density:0.69 g/cm3 (15 °C)
Storage Temperature:Ambient
EINECS:205-563-8
UN:1206
ADR:3,II
 :GHS09
 :GHS07
 :GHS08
 :GHS02
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544