Order Entry
Belgium
ContactUsLinkComponent
Methanol, Extra pure meets analytical specifications of Reagent Ph. Eur. acetone-free, Burdick & Jackson™
Health
Toxic
Flammable
Methanol, Extra pure meets analytical specifications of Reagent Ph. Eur. acetone-free, Burdick & Jackson™
Catalog # HONC65805-200L
   Safer Human and Environmental Health
CAS Number:  
Health
Toxic
Flammable
Methanol, Extra pure meets analytical specifications of Reagent Ph. Eur. acetone-free, Burdick & Jackson™
Catalog # HONC65805-200L
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Metal drum
  • Pk:
    200 L
  • Seq:
    5549
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.7 %
  • Boiling range:
    64 - 65 °C
  • Density (D 20/20):
    0.791 - 0.793
  • Refractive index (n 20/D):
    1.328 - 1.330
  • Non-volatile matter:
    Max. 10 ppm
  • Water (Karl Fischer):
    Max. 0.10 %
  • Free acid (as HCOOH):
    Max. 0.003 %
  • Acidity or alkalinity:
    complying
  • Iron (Fe):
    Max. 0.0001 %
  • Chloride (Cl):
    Max. 0.0001 %
  • Sulfate (SO₄):
    Max. 0.0005 %
  • Acetone (GC):
    Max. 0.001 %
  • Benzene (GC):
    Max. 2 ppm
  • Reaction against H₂SO₄:
    complying
  • Reducing impurities:
    complying
  • Identity (IR):
    complying
  • UV-absorption:
    complying
  • Impurity A (GC):
    complying
  • Related substances (GC):
    complying
  • Appearance:
    complying
  • Residual solvents:
    complying

Specification Test Results

About this item

 :
 :32.04 g/mol
Boiling Pt:64…65 °C
Density:0.79 g/cm3
Flash Pt:11 °C
UN:1230
ADR:3,II
 :GHS08
 :GHS06
 :GHS02
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595