Order Entry
Belgium
ContactUsLinkComponent
n-Hexane for gas chromatography, Burdick & Jackson™
n-Hexane for gas chromatography, Burdick & Jackson™
Certificates
undefined
n-Hexane for gas chromatography, Burdick & Jackson™

Specification Test Results

  • Assay (GC):
    Min. 95.0 %
  • Non-volatile matter:
    Max. 0.0003 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Suitability for residue analysis:
    Max. 5 ng/l

Specification Test Results

Frequently Bought Together