Order Entry
Belgium
ContactUsLinkComponent
n-Hexane for HPLC, Burdick & Jackson™
n-Hexane for HPLC, Burdick & Jackson™
Catalog # HONC65730-2.5L
CAS Number:  
undefined
n-Hexane for HPLC, Burdick & Jackson™
Catalog # HONC65730-2.5L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (GC):
    Min. 97 %
  • Non-volatile matter:
    Max. 0.0003 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Acidity:
    Max. 0.0001 meq/g
  • Alkalinity:
    Max. 0.0002 meq/g
  • Transmittance at 200 nm:
    Min. 30 %
  • Transmittance at 225 nm:
    Min. 90 %
  • Transmittance from 250 nm:
    Min. 99 %
  • Phthalates as dibutylphthalate:
    Max. 0.1 mg/l

Specification Test Results

About this item

 :
 :86.18 g/mol
Boiling Pt:66…69 °C (1.013 hPa)
Melting Pt:–95 °C
Density:0.660…0.668 g/cm3 (20 °C)
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520