


Methanol Reag. Ph. Eur. for HPLC, Burdick & Jackson™
HONC65725-200L-RC
:
- Pack type:Stainless steel drum, returnable
- Pk:200 L
- Seq:5549
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Appearance:complying
- Assay (GC):Min. 99.9 %
- Boiling range:64 - 65 °C
- Density (D 20/20):0.791 - 0.793
- Non-volatile matter:Max. 0.0003 %
- Water (Karl Fischer):Max. 0.02 %
- Acidity:Max. 0.0002 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Absorbance at 210 nm:Max. 0.70
- Absorbance at 220 nm:Max. 0.30
- Absorbance at 225 nm:Max. 0.17
- Absorbance at 230 nm:Max. 0.13
- Absorbance at 250 nm:Max. 0.02
- Absorbance at 260 nm:Max. 0.01
- Fluorescence (chinin) at 254 nm:Max. 1 ppb
- Fluorescence (chinin) at 365 nm:Max. 1 ppb
- Phthalates as dibutylphthalate:Max. 0.1 mg/l
:
:32.04 g/mol
:64…65 °C
:0.79 g/cm3
:11 °C
:1230
:3,II
:GHS06
:GHS08
:GHS02
:67-56-1
:CH₃OH
:MFCD00004595