Order Entry
Belgium
ContactUsLinkComponent
n-Hexane, dried (max. 10 ppm H₂O), Burdick & Jackson™
n-Hexane, dried (max. 10 ppm H₂O), Burdick & Jackson™
Catalog # HONC65519-185L
CAS Number:  
n-Hexane, dried (max. 10 ppm H₂O), Burdick & Jackson™
Catalog # HONC65519-185L
CAS Number:  

Specifications

  • Pack type:
    Stainless steel drum
  • Pk:
    185 L
  • Seq:
    5491

Specifications

Specification Test Results

  • Assay of hexane (all isomers, GC):
    Min. 99.0 %
  • Assay of n-Hexane (GC):
    Min. 95.0 %
  • APHA:
    Max. 10
  • Density (D 20/4):
    0.658 - 0.662
  • Refractive index (n 20/D):
    1.374 - 1.376
  • Water (Karl Fischer):
    Max. 100 ppm

Specification Test Results

About this item

 :
 :86.18 g/mol
Boiling Pt:66…69 °C
Density:0.660…0.668 g/cm3
Flash Pt:–26 °C
UN:1208
ADR:3,II
 :GHS08
 :GHS07
 :GHS09
 :GHS02
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520