Isopropyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:108-21-4
:
Formula:C₅H₁₀O₂
MDL Number:MFCD00008877
:102.13 g/mol
Merck Index:13,05225
Boiling Pt:85 °C (1013 hPa)
Melting Pt:–62 °C
Density:0.8732 g/cm³ (20 °C)
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-561-1
UN:1220
ADR:3,II
:GHS02
:GHS07
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Assay:Min. 99 %
- Identification (IR):Passes test
- Density (20/4):0.871 --> 0.873
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC87068.290 spec_VWRC87068.320 spec_VWRC87068.320
Product specification
Frequently Bought Together