Order Entry
Belgium
Orders LinkContactUsLinkComponent
Inoculation loop holders

Inoculation loop holders

Inoculation loop holder, 215 mm
  WART6.000.365
 :   WARTEWIG
Inoculation loop holders
  WART6.000.365
 :  WARTEWIG
Inoculation loop holders

Inoculation loop holder, 215 mm

 
66,30 €
   
1 * 4 ST
 

These inoculation loop holders are made from stainless steel.

  • With sleeve nut, colour coded
  • For inoculation loops with wire Ø 0,6 to 1 mm
  • Autoclavable
 
  • Description:
    Inoculation loop holder
  • Length:
    215 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.