ContactUsLinkComponent
J.T.Baker® BAKERBOND® PROchievA™, Affinity Chromatography Columns, Recombinant Protein A Resin
 :  Avantor
undefined
J.T.Baker® BAKERBOND® PROchievA™, Affinity Chromatography Columns, Recombinant Protein A Resin
 :  Avantor
  • Catalog No:
  • C789-25
  • C789-07
  • C789-11
  • C789-18
  • Binding capacity:
  • FIO (Target > 65 mg/ml)
  • FIO (Target > 65 mg/ml)
  • FIO (Target > 65 mg/ml)
  • FIO (Target > 65 mg/ml)
  • Storage:
  • +2...+8 °C
  • +2...+8 °C
  • +2...+8 °C
  • +2...+8 °C
Frequently Bought Together