Orders LinkContactUsLinkComponent
Measuring spoons, blue
Measuring spoons, blue
Supplier:   BURKLE
Measuring spoons, blue
Supplier:  BURKLE
Certificates
Measuring spoons, blue
Measuring spoons, blue
16 Options Available Below
Product Details & Documents
The shade of blue that is used does not occur in a natural form in foodstuffs. Because of this, blue products or parts of them can be quickly found and easily identified in a visual check if they are ever mixed into production.
  • PS, blue
  • Can be used as part of HACCP/IFS foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
  • With stripper edge
  • For single use
  • Clean room manufactured (clean room class 7)
The use of blue tools thus helps to meet the requirements of the HACCP hazard analysis and assessment in reference to foreign objects. This is an important criterion, especially in food processing and production and in the animal feed and pharmaceutical industry.
Volumetric and dosing spoons can be used to measure small amounts of powder, granulates and liquids exactly and dose them precisely. For precise measuring, excess medium is stripped off the upper edge of the spoon. The content volume is engraved on the handle.
The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom of the container or for getting into the corners of the container, thus allowing for good emptying of small remaining amounts. The flat base makes it possible to set the volumetric spoon down without spilling the content.
The disposable dosing spoon is suitable for use as a sample spoon for taking small sample quantities. Since the plastic volumetric spoon was developed for single use, there is no cleaning and the possibility of cross-contamination and impurities is excluded.
The dosing spoons and volumetric spoons are suitable for precise measuring for chemical and pharmaceutical applications, cosmetics, animal feed and the food & beverage industry.
Certifications: Complies with EU food requirements and FDA regulations.
Ordering information: Available either in a large package with 100 pieces or 100 pieces packaged individually and radiation sterilised.
Product Family Options
Non sterile
Product Information
  • Capacity
    Availability
    Price
  • 300-0387
    Large package
    Large package
    10,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 51,50 €
    51,50 €
    1 * 100 ST
       51,50 €
  • 300-0389
    Large package
    Large package
    15,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 54,90 €
    54,90 €
    1 * 100 ST
       54,90 €
  • 300-0379
    Large package
    Large package
    0,5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 35,70 €
    35,70 €
    1 * 100 ST
       35,70 €
  • 300-0393
    Large package
    Large package
    50,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 90,30 €
    90,30 €
    1 * 100 ST
       90,30 €
  • 300-0381
    Large package
    Large package
    1,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 39,30 €
    39,30 €
    1 * 100 ST
       39,30 €
  • 300-0391
    Large package
    Large package
    25,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 63,00 €
    63,00 €
    1 * 100 ST
       63,00 €
  • 300-0383
    Large package
    Large package
    2,5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 43,50 €
    43,50 €
    1 * 100 ST
       43,50 €
  • 300-0385
    Large package
    Large package
    5,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 47,60 €
    47,60 €
    1 * 100 ST
       47,60 €
Sterile
Product Information
  • Capacity
    Availability
    Price
  • 300-0386
    Individually packaged
    Individually packaged
    5,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 83,90 €
    83,90 €
    1 * 100 ST
       83,90 €
  • 300-0388
    Individually packaged
    Individually packaged
    10,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 88,20 €
    88,20 €
    1 * 100 ST
       88,20 €
  • 300-0394
    Individually packaged
    Individually packaged
    50,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 177,00 €
    177,00 €
    1 * 100 ST
       177,00 €
  • 300-0380
    Individually packaged
    Individually packaged
    0,5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 69,70 €
    69,70 €
    1 * 100 ST
       69,70 €
  • 300-0382
    Individually packaged
    Individually packaged
    1,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 73,60 €
    73,60 €
    1 * 100 ST
       73,60 €
  • 300-0390
    Individually packaged
    Individually packaged
    15,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 92,40 €
    92,40 €
    1 * 100 ST
       92,40 €
  • 300-0392
    Individually packaged
    Individually packaged
    25,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 108,00 €
    108,00 €
    1 * 100 ST
       108,00 €
  • 300-0384
    Individually packaged
    Individually packaged
    2,5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 77,70 €
    77,70 €
    1 * 100 ST
       77,70 €