Orders LinkContactUsLinkComponent
Dosing spoons, white
Supplier:   BURKLE
Dosing spoons, white
Supplier:  BURKLE
Certificates
Dosing spoons, white
Dosing spoons, white
16 Options Available Below
Product Details & Documents
Volumetric and dosing spoons can be used to measure small amounts of powder, granulates and liquids exactly and dose them precisely. For precise measuring, excess medium is stripped off the upper edge of the spoon. The content volume is engraved on the handle.
  • PS, white
  • Optionally packed individually and sterilised
  • Stripper edge
  • For single use
  • Cleanroom manufactured (cleanroom Class 7)
The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom or getting into the corners of a container, thus allowing for efficient emptying of small remaining amounts. The flat base makes it possible to set the volumetric spoon down without spilling the contents.
The disposable dosing spoon is suitable for use as a sample spoon for taking small sample quantities. Since this plastic volumetric spoon was developed for single use, there is no cleaning, and the possibility of cross-contamination and impurities is excluded.
These dosing and volumetric spoons are suitable for precise measurements in chemical and pharmaceutical applications, cosmetics, animal feed also the food and beverage industry.
 : Complies with EU food requirements and FDA regulations.
 : Available either in a large pack of 100 pieces or 100 pieces packed individually and radiation sterilised.
Product Family Options
Non sterile
Product Information
  • Capacity
    Availability
    Price
  • 300-0372
    Large package
    Large package
    15,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 54,90 €
    54,90 €
    1 * 100 ST
       54,90 €
  • 300-0364
    Large package
    Large package
    1,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 39,30 €
    39,30 €
    1 * 100 ST
       39,30 €
  • 300-0370
    Large package
    Large package
    10,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 51,50 €
    51,50 €
    1 * 100 ST
       51,50 €
  • 300-0368
    Large package
    Large package
    5,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 47,60 €
    47,60 €
    1 * 100 ST
       47,60 €
  • 300-0374
    Large package
    Large package
    25,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 63,00 €
    63,00 €
    1 * 100 ST
       63,00 €
  • 300-0376
    Large package
    Large package
    50,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 90,30 €
    90,30 €
    1 * 100 ST
       90,30 €
  • 300-0362
    Large package
    Large package
    0,5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 35,70 €
    35,70 €
    1 * 100 ST
       35,70 €
  • 300-0366
    Large package
    Large package
    2,5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 43,50 €
    43,50 €
    1 * 100 ST
       43,50 €
Sterile
Product Information
  • Capacity
    Availability
    Price
  • 300-0373
    Packaged individually
    Packaged individually
    15,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 92,40 €
    92,40 €
    1 * 100 ST
       92,40 €
  • 300-0375
    Packaged individually
    Packaged individually
    25,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 108,00 €
    108,00 €
    1 * 100 ST
       108,00 €
  • 300-0377
    Packaged individually
    Packaged individually
    50,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 177,00 €
    177,00 €
    1 * 100 ST
       177,00 €
  • 300-0365
    Packaged individually
    Packaged individually
    1,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 73,60 €
    73,60 €
    1 * 100 ST
       73,60 €
  • 300-0369
    Packaged individually
    Packaged individually
    5,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 83,90 €
    83,90 €
    1 * 100 ST
       83,90 €
  • 300-0371
    Packaged individually
    Packaged individually
    10,0 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 88,20 €
    88,20 €
    1 * 100 ST
       88,20 €
  • 300-0367
    Packaged individually
    Packaged individually
    2,5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 77,70 €
    77,70 €
    1 * 100 ST
       77,70 €
  • 300-0363
    Packaged individually
    Packaged individually
    0,5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 69,70 €
    69,70 €
    1 * 100 ST
       69,70 €