Order Entry
Belgium
ContactUsLinkComponent
N-(Propionyloxy)succinimide ≥95% (by HPLC), LiChropur™ derivatisation reagent, for LC-MS, Supelco®
N-(Propionyloxy)succinimide ≥95% (by HPLC), LiChropur™ derivatisation reagent, for LC-MS, Supelco®
Catalog # 93535-1G
Supplier:  Merck
CAS Number:  
N-(Propionyloxy)succinimide ≥95% (by HPLC), LiChropur™ derivatisation reagent, for LC-MS, Supelco®
Catalog # 93535-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance (Color):
    White to Faint Beige
  • Appearance (Form):
    Powder or Crystals
  • Purity (HPLC Area %):
    ≥ 95 %
  • Infrared Spectrum:
    Conforms to Structure

Specification Test Results

About this item

Synonyms: 1-(1-Oxopropoxy)-2,5-pyrrolidinedione, 2,5-Dioxopyrrolidin-1-yl-propanoate, Propanoic acid-NHS ester, Succinimidyl propionate, Propionyl-Osu
 :
 :171.15 g/mol
Melting Pt:44...48 °C
 :GHS07
CAS Number:30364-55-7
Formula:C₇H₉NO₄
MDL Number:MFCD03427483