Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 1 M for HPCE, Supelco®
Sodium hydroxide 1 M for HPCE, Supelco®
Supplier:  Merck
Certificates
Sodium hydroxide 1 M for HPCE, Supelco®
Supplier:  Merck

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance (Color):
    Colorless
  • Appearance (form):
    Liquid
  • Titration (Molarity):
    0.9 - 1.1 mol/l
  • UV-Absorption Values:
    260nm ≤ 0.5, 270nm ≤ 0.2
  • Test by UV Spectroscopy:
    280nm ≤ 0.03, 290nm ≤ 0.01, 500nm ≤ 0.01
  • Luminescence:
    Corresponds to Requirements
  • Residue (Filter Test):
    No Residue
  • Filtertest (Method):
    Neat
  • Recommended Retest Period:
    84 Months

Specification Test Results

Frequently Bought Together