DEV gelatin agar medium, Millipore®
Catalog # 1.10685.0500
Supplier: Merck
CAS Number:
Specifications
- Pk:500 g
- Standards:DIN EN ISO 9308-1
- Description:DEV gelatine agar
- Environmentally Preferable:
- Storage temperature:15...25 °C
- pH:7,1 - 7,5 (50 g/l, H₂O, 22 °C)
- Physical form:Liquid
Specifications
About this item
DEV gelatin agar for microbiology.