Orders LinkContactUsLinkComponent
Combs for gel casting system, MultiCaster
Supplier:   EMBITEC
Combs for gel casting system, MultiCaster
Supplier:  EMBITEC
Certificates
Combs for gel casting system, MultiCaster
3 Options Available Below
Product Details & Documents
Reversible combs for the MultiCaster gel casting system with dual configuration for multiple gel formats.
  • Made of polycarbonate
  • Well configurations and estimated sample volume: 12+1 well (30 μl) and 24+1 well (11 μl)
  • Multi-channel pipette compatibility: 12-channel
  • Estimated sample volume is based on gel thickness of 6,5 mm
All combs are interchangeable so that a single MultiCaster system can be used for all preparations and high throughput needs by simply swapping the combs.
Product Family Options
Product Information
  • Availability
    Price
  • 730-0361
    Multicaster comb, aqua
    Multicaster comb, aqua
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 97,80 €
    97,80 €
    1 * 2 ST
       97,80 €
  • 730-0362
    Multicaster comb, blue
    Multicaster comb, blue
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 97,80 €
    97,80 €
    1 * 2 ST
       97,80 €
  • 730-0363
    Multicaster comb, orange
    Multicaster comb, orange
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 97,80 €
    97,80 €
    1 * 2 ST
       97,80 €