Orders LinkContactUsLinkComponent
Ground socket
Ground socket
NS 29/32
Catalog # BRND51467
Supplier:   Brand
Ground socket
Catalog # BRND51467
Supplier:  Brand
Ground socket
Ground socket
NS 29/32
Price
29,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
PTFE. With grip collar and sealing rings for excellent tightness.
  • Greaseless seal will not contaminate sample
  • Outstanding chemical resistance at operating temperatures from −200 to +260 °C
  • Sturdy design for permanent use
Low leakage rate: Q < 1 · 10-4 mbar · l · s-1.
Specifications
  • For:
    NS 29/32