Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Media for Microbiology
Prepared Media for Microbiology
VWR®, Demi-Fraser broth tubes
VWR®, Demi-Fraser broth tubes
VWR®, Demi-Fraser broth tubes
Catalog # 5004.00010
Supplier:
BALIS
CAS Number:
Expand image
VWR®, Demi-Fraser broth tubes
Catalog # 5004.00010
Supplier:
BALIS
CAS Number:
Specifications
Pk:
100
Description:
Demi Fraser broth, glass/PP cap
Volume:
10 ml
Specifications
About this item
Product Details
Product Details