Coated filter, ORBO™ 80
37 mm
Catalog # 20811.
Supplier: Merck
Jump to:
Product Details & Documents
ORBO™ 80 is a glass fiber filter coated with 1-(2-pyridyl)piperazine.
- No cassettes
It is used for sampling diisocyanates in air.
Specifications
- Size:37 mm