Order Entry
Belgium
ContactUsLinkComponent
Ethanol absolute (untaxed) ≥99.8% (by GC), SPECTRANAL™, Riedel-de Haën™
Irritant
Flammable
Ethanol absolute (untaxed) ≥99.8% (by GC), SPECTRANAL™, Riedel-de Haën™
Catalog # 34935-1L
CAS Number:  
Irritant
Flammable
Ethanol absolute (untaxed) ≥99.8% (by GC), SPECTRANAL™, Riedel-de Haën™
Catalog # 34935-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.8 Vol.%
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.2 %
  • Free acid (as CH₃COOH):
    Max. 0.001 %
  • Suitability for UV spectroscopy:
    complying
  • Suitability for IR spectroscopy:
    complying
  • Transmittance at 210 nm:
    Min. 20 %
  • Transmittance at 220 nm:
    Min. 40 %
  • Transmittance at 230 nm:
    Min. 60 %
  • Transmittance at 240 nm:
    Min. 80 %
  • Transmittance from 260 nm:
    Min. 98 %
  • Fluorescence (chinin) at 254 nm:
    Max. 1 ppb
  • Fluorescence (chinin) at 365 nm:
    Max. 1 ppb
  • Autoignition Temperature:
    683 °F
  • Vapor Pressure:
    44.6 mmHg ( 20 °C)

Specification Test Results

About this item

 :
 :46.07 g/mol
Boiling Pt:78…79 °C (1.013 hPa)
Melting Pt:–115 °C
Density:0.790…0.791 g/cm3 (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
 :GHS07
 :GHS02
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568