ContactUsLinkComponent
Drop-out mix synthetic minus adenine, histidine, leucine, tryptophan, uracil without yeast nitrogen base
Drop-out mix synthetic minus adenine, histidine, leucine, tryptophan, uracil without yeast nitrogen base
Catalog # USBID9540-05
Supplier:  US Biological
CAS Number:  
Drop-out mix synthetic minus adenine, histidine, leucine, tryptophan, uracil without yeast nitrogen base
Catalog # USBID9540-05
Supplier:  US Biological
CAS Number:  
Specifications
  • Pk:
    100 g
  • Description:
    D9540 Drop-out Mix Synthetic Minus Histidine, Leucine, Tryptophan, Uracil without Yeast Nitrogen Base (DO, Dropout, Powder)
Specifications
About this item
Drop-out mixes are used as a supplement to Drop-out base for making synthetic, complete media formulations in accordance with methods in yeast genetics.
  • Convenient powders
  • Useful for one-hybrid and two-hybrid systems
All lots are quality control tested using the appropriate yeast strains and transformants.