Order Entry
Belgium
ContactUsLinkComponent
n-Decane ≥99%, GPR RECTAPUR®
n-Decane ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
n-Decane ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Boiling point:
    Min. 170 °C
  • Density (20/4):
    0,729 --> 0,730
  • Aromatics:
    Max. 0,02 %

Specification Test Results

Frequently Bought Together