Order Entry
Belgium
ContactUsLinkComponent
 
1,2,4-Trichlorobenzene ≥98%, GPR RECTAPUR®
  85997.290
 :  
undefined
1,2,4-Trichlorobenzene ≥98%, GPR RECTAPUR®
  85997.290
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

 

 

  • Assay:
    Min 98%
  • Identification (IR):
    Passes test
  • Density (20/4):
    1,454 - 1,455
  • Refractive index (20 °C, 589 nm):
    1,570 - 1,572

 

 

 :
 :181.45 g/mol
 :17 °C
 :204-428-0
 :2321
 :GHS07
 :GHS09
 :Ambient
 :120-82-1
 :C₆H₃Cl₃
 :MFCD00000547
 :214.4 °C (1013 hPa)
 :1.45 g/cm³ (20 °C)
 :99 °C
 :6.1 III
 :13,09704