Phenol red solution pH-indicator (pH 6,5 - 8,5)
: Hach
:143-74-8
:
:C₁₉H₁₄O₅S
:MFCD00003552
:354.38 g/mol
Frequently Bought Together
pH buffers, ready to use
: WTW
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Decontamination Solution, DNA/RNA-ExitusPlus™
: PanReac AppliChem
VWR®, Pipette Tips, Universal
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Titrants and reagents for volumetric Karl Fischer titrations, HYDRANAL™, Fluka™
: Honeywell Chemicals
VWR®, Accessories for VWR Heating and Cooling Circulators
: VWR Collection
Starch, soluble 1% (w/w) in aqueous solution stabilised, pure
: Thermo Fisher Scientific