Indicator buffer tablets for the determination of water hardness with Titriplex® solutions, Supelco®
Supplier: Merck
Certificates
About this item
- Tablet disintegration time (at 25 °C) ≤ 150 s
- Tablet weight: 0,20 g ≤ w ≤ 0,23 g