Order Entry
Belgium
ContactUsLinkComponent
Lectin from Triticum vulgaris, immobilized on Agarose CL-4B, for affinity chromatography, Supelco®
Lectin from Triticum vulgaris, immobilized on Agarose CL-4B, for affinity chromatography, Supelco®
  61768-1ML
 :  Merck
 :  
Lectin from Triticum vulgaris, immobilized on Agarose CL-4B, for affinity chromatography, Supelco®
  61768-1ML
 :  Merck
 :  

 

  • Particle Size:
    45 - 165 µm
  • Pk:
    1 mL
  • Pore size:
    50 - 160 µm
  • Description:
    Lectin from Triticum vulgaris, immobilized on Agarose CL-4B, 50% aqueous suspension in 1 M NaCl

 

 

50% aqueous suspension in 1 M NaCl.

  • Capacity: 0,1 to 1,0 mg/ml, packed gel capacity (ovomucoid)

Application: Protein isolation from bordetella pertussis that facilitates entry of calmodulin-sensitive adenylate cyclase into animal cells.

 : Storage temperature: Refrigerator.