ContactUsLinkComponent
ISA (ionic strength adjustment solution: 1 M KCl), Supelco®
ISA (ionic strength adjustment solution: 1 M KCl), Supelco®
Catalog # 58221-500ML-F
Supplier:  Merck
CAS Number:  
ISA (ionic strength adjustment solution: 1 M KCl), Supelco®
Catalog # 58221-500ML-F
Supplier:  Merck
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    ISA (ionic strength adjustment solution)
  • M-Clarity quality level:
    MQ100
  • Contents:
    1 M KCl
Specifications
About this item
Solution used to establish a constant ionic strength in a sample solution containing e.g. calcium for det. with ISE.
  • Prepared with high purity salts