Order Entry
Belgium
ContactUsLinkComponent
 
Tris-EDTA (TE) buffer solution for molecular biology, pH 7,4, BioUltra, Supelco®
  93302-100ML
 :  Merck
 :  
undefined
Tris-EDTA (TE) buffer solution for molecular biology, pH 7,4, BioUltra, Supelco®
  93302-100ML
 :  Merck
 :  

 

  • Concentration:
    10 mM Tris-HCl; 1 mM EDTA
  • Pk:
    100 mL
  • Description:
    Tris-EDTA buffer solution
  • Seq:
    0001
  • M-Clarity quality level:
    MQ100
  • pH accuracy:
    ±0,2
  • pH value:
    7,4

 

 

Tris-EDTA buffer solution has been used for the dilution of DNA samples.[1] It has been used for the elution of purified vector.