Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffers, colour coded, reference standards, Supelco®

pH buffers, colour coded, reference standards, Supelco®

Supplier:   Merck
pH buffers, colour coded, reference standards, Supelco®
Supplier:  Merck
Certificates
pH buffers, colour coded, reference standards, Supelco®
pH buffers, colour coded, reference standards, Supelco®

7 Options Available Below

Product Details & Documents

Certifications: Standardised against N.I.S.T. standard reference materials

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • B5020-500ML
  • B5020-6X500ML
  • B5020-1GA
  • B4770-500ML
  • B4770-1GA
  • B4895-500ML
  • B4895-6X500ML
  • M-Clarity quality level:
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100

Specifications

Product Family Options

Product Information

  • pH value
    pH accuracy
    Colour
    Reference temperature
    Pack type
    Pk Info
    Pk
    Availability
    Price
  • 4,00
    ±0,01
    Red
    25 °C
    Plastic bottle
    1×500 ml
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 49,60 €
    49,60 €
    1 * 500 mL
       49,60 €

    Specifications:
    More
    Specifications
    Cat. No.
    B5020-500ML
    M-Clarity quality level
    MQ100
  • 4,00
    ±0,01
    Red
    25 °C
    Plastic bottle
    6×500 ml
    6
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 233,00 €
    233,00 €
    1 * 6 ST
       233,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    B5020-6X500ML
    M-Clarity quality level
    MQ100
  • 4,00
    ±0,01
    Red
    25 °C
    Plastic container with spigot
    1×gallon
    1 gal US
    Log in for availability
    Log in for availability
    1 * 1 gal us
    1 * 1 gal us - 233,00 €
    233,00 €
    1 * 1 gal us
       233,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    B5020-1GA
    M-Clarity quality level
    MQ100
  • 7,00
    ±0,01
    Yellow
    25 °C
    Plastic bottle
    1×500 ml
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 49,60 €
    49,60 €
    1 * 500 mL
       49,60 €

    Specifications:
    More
    Specifications
    Cat. No.
    B4770-500ML
    M-Clarity quality level
    MQ100
  • 7,00
    ±0,01
    Yellow
    25 °C
    Plastic container with spigot
    1×gallon
    1 gal US
    Log in for availability
    Log in for availability
    1 * 1 gal us
    1 * 1 gal us - 259,00 €
    259,00 €
    1 * 1 gal us
       259,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    B4770-1GA
    M-Clarity quality level
    MQ100
  • 10,00
    ±0,01
    Blue
    25 °C
    Plastic bottle
    1×500 ml
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 49,60 €
    49,60 €
    1 * 500 mL
       49,60 €

    Specifications:
    More
    Specifications
    Cat. No.
    B4895-500ML
    M-Clarity quality level
    MQ100
  • 10,00
    ±0,01
    Blue
    25 °C
    Plastic bottle
    6×500 ml
    6
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 231,00 €
    231,00 €
    1 * 6 ST
       231,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    B4895-6X500ML
    M-Clarity quality level
    MQ100