Order Entry
Belgium
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Phosphorus standard solution
  P3869-25ML
 :  Merck
 :  
Supelco® Organic Reference Standard, Phosphorus standard solution
  P3869-25ML
 :  Merck
 :  

 

  • Concentration:
    0,65 mM (phosphorous)
  • Pk:
    25 mL
  • Description:
    Phosphorus standard solution
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H290
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100

 

 

  • GHS signal: GHS05
 : P234 - P390