Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, Glucose Standard Solution
Supelco® Organic Reference Standard, Glucose Standard Solution
Catalog # G6918-100ML
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Glucose Standard Solution
Catalog # G6918-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1 mg/ml
  • Pk:
    100 mL
  • Description:
    Glucose standard solution
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100

Specifications

About this item