Orders LinkContactUsLinkComponent
MS calibration solution, API, sodium iodide and cesium iodide, Supelco®
Sodium iodide (2 μg/μL) and cesium iodide (50 ng/μL) in 50/50 2-propanol/water, 30 mL
Catalog # 14379-30ML
Supplier:   Merck
MS calibration solution, API, sodium iodide and cesium iodide, Supelco®
Catalog # 14379-30ML
Supplier:  Merck
CAS Number:  
MS calibration solution, API, sodium iodide and cesium iodide, Supelco®
Sodium iodide (2 μg/μL) and cesium iodide (50 ng/μL) in 50/50 2-propanol/water, 30 mL
CAS Number:  
Price
92,30 €
Log in for availability
1 * 30 mL
Product Details & Documents
Solution for LC-MS calibration.
Documents
Specifications
Specifications
  • Pk:
    30 mL
  • Description:
    Sodium iodide (2 μg/μL) and cesium iodide (50 ng/μL) in 50/50 2-propanol/water