N,N-Dimethyl-p-toluidine ≥98.0% (by GC), GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:99-97-8
:
Formula:C₉H₁₃N
MDL Number:MFCD00008316
:135.21 g/mol
UN:2810
ADR:6.1,III
:GHS06
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear pale yellow to amber liquid
- Assay (GC):min. 98,0 %
- Water:max. 0,5 %
- Density (d20°C/4°C):0,935 – 0,937
- Identity (IR):Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC85986.230 spec_VWRC85986.290 spec_VWRC85986.460
Product specification
Frequently Bought Together