Order Entry
Belgium
ContactUsLinkComponent
Sodium-3-[ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulphonate hydrate ≥98.0% (dried basis, T) for spectrophotometric determination H₂O₂, Supelco®
Sodium-3-[ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulphonate hydrate ≥98.0% (dried basis, T) for spectrophotometric determination H₂O₂, Supelco®
Catalog # 04340-500MG
Supplier:  Merck
CAS Number:  
undefined
Sodium-3-[ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulphonate hydrate ≥98.0% (dried basis, T) for spectrophotometric determination H₂O₂, Supelco®
Catalog # 04340-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :295.33 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:82692-93-1
Formula:C₁₂H₁₉NO₄S
MDL Number:MFCD00040641