Order Entry
Belgium
ContactUsLinkComponent
Sodium polytungstate ~85% in water for separation of minerals mixtures, Supelco®
Sodium polytungstate ~85% in water for separation of minerals mixtures, Supelco®
Catalog # 80656-100ML
Supplier:  Merck
CAS Number:  
undefined
Sodium polytungstate ~85% in water for separation of minerals mixtures, Supelco®
Catalog # 80656-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Sodium polytungstate monohydrate, Sodium tungstate (meta) monohydrate, hexa-Sodium tungstate (W12) monohydrate
 :
 :2986.13 g/mol (2968.12 anhy)
 :GHS05
CAS Number:12141-67-2
Formula:Na₆O₃₉W₁₂·H₂O
MDL Number:MFCD00061466