8-Hydroxy-7-iodoquinoline-5-sulphonic acid ≥98.5% for spectrophotometric determination of Fe, Supelco®
Catalog # 55370-100G
Supplier: Merck
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:351.11 g/mol
ADR:8,III
:M-Clarity™ quality level = MQ100
UN:2585
:GHS05
CAS Number:547-91-1
Formula:C₉H₆INO₄S
MDL Number:MFCD00006793