Order Entry
Belgium
ContactUsLinkComponent
Nitric acid 25%, Titripur® for argentometric titration, Supelco®
Nitric acid 25%, Titripur® for argentometric titration, Supelco®
Catalog # 1.60317.0500
Supplier:  Merck
CAS Number:  
Nitric acid 25%, Titripur® for argentometric titration, Supelco®
Catalog # 1.60317.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    0,5 L
  • Seq:
    5036
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Nitric acid solution
 :
 :63.01 g/mol
ADR:8,
 :M-Clarity™ quality level = MQ100
UN:2031
 :GHS05
Melting Pt:-42
Storage Temperature:Ambient
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349
Boiling Pt:87
Density:1.51
ADR:8
Merck Index:13,06610