ContactUsLinkComponent
Sterile sleeve/glove system, BioClean™
Sterile sleeve/glove system, BioClean™
Supplier:  Nitritex
Certificates
undefined
Sterile sleeve/glove system, BioClean™
Supplier:  Nitritex
Specifications
  • Catalog No:
  • 112-6122
  • 112-6123
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Cleanroom class:
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • Material:
  • Nitrile
  • Nitrile
  • Finish:
  • Smooth
  • Smooth
  • Cuff style:
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,11 mm
  • 0,11 mm
  • Finger thickness:
  • 0,18 mm
  • 0,18 mm
  • Palm thickness:
  • 0,15 mm
  • 0,15 mm
  • EN norm:
  • EN ISO 374-1 Type B (KOT), EN ISO 374-5 VIRUS, EN 421
  • EN ISO 374-1 Type B (KOT), EN ISO 374-5 VIRUS, EN 421
  • Sterility:
  • Sterile
  • Sterile