

Paraformaldehyde 4% (w/v) in aqueous solution methanol-free
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:50-00-0
:
Formula:CH₂O
MDL Number:MFCD00003274
:30.03 g/mol
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Concentration (w/v):3.5 to 4%
- Appearance:Clear colourless liquid
- Ash:<=0.01% (Paraformaldehyde content)
- Iron (Fe):<=2.0% (Paraformaldehyde content)
Specification Test Results
Frequently Bought Together