Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Filters
Precolumn Filters and Frits
Avantor® Hichrom, Preparative Frit Assembly
Avantor® Hichrom, Preparative Frit Assembly
Avantor® Hichrom, Preparative Frit Assembly
Catalog # HICHHI-140
Supplier:
Avantor
Expand image
Avantor® Hichrom, Preparative Frit Assembly
Catalog # HICHHI-140
Supplier:
Avantor
Specifications
Porosity:
2,0 µm
Colour:
Natural
Description:
Preparative frit assembly
Pk:
10
Specifications
About this item
Product Details
Product Details