Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

TLC developing chambers

Separating chambers for TLC micro-sets, 63×109 mm
  552-0273
TLC developing chambers
  552-0273
TLC developing chambers
TLC developing chambers

Separating chambers for TLC micro-sets, 63×109 mm

 
41,70 €
   
1 * 4 ST
 

Mason jar shaped developing chambers for TLC application. Glass of hydrolytic category III, lid is made of PP with LDPE sealing disk.

Recommended TLC plate size: Up to 4×8 cm

 
  • Description:
    Separating chambers for TLC micro-sets
  • Seq:
    03
  • Ø×H:
    63×109 mm