ContactUsLinkComponent
Avantor® Hichrom, UHPLC Column Connector
Avantor® Hichrom, UHPLC Column Connector
Catalog # HICHHI-903
Supplier:  Avantor
undefined
Avantor® Hichrom, UHPLC Column Connector
Catalog # HICHHI-903
Supplier:  Avantor
Specifications
  • Description:
    UHPLC connector for 1/16" O.Ø tubing
  • Pk:
    1
Specifications
About this item
This reusable UHPLC column connector have 10-32 threads. This fitting is reusable, as they are not permanently swaged onto the inlet tubing.
  • Compatible with all UHPLC systems and column brands
  • Avoids poor connections
  • Pressure rated to >1700 bar (>25000 psi)
An internal ferrule design prevents tubing slippage, whilst the PEEK-polymer blend front ferrule eliminates the risk of damage to receiving ports.