Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tubing
Chromatography Tubing
Avantor® Hichrom PEEKsil™, Tubing Sleeve, PEEK
Avantor® Hichrom PEEKsil™, Tubing Sleeve, PEEK
Avantor® Hichrom PEEKsil™, Tubing Sleeve, PEEK
Catalog # HICHHI-1113
Supplier:
Avantor
Expand image
Avantor® Hichrom PEEKsil™, Tubing Sleeve, PEEK
Catalog # HICHHI-1113
Supplier:
Avantor
Specifications
Ext.Ø:
1,575 mm
Ø int.:
0,864 mm
Colour:
Blue
Pk:
1
Specifications
About this item
Product Details
Product Details