Order Entry
Belgium
ContactUsLinkComponent
Peqlab peqGOLD, Plasmid Purification, Plasmid Miniprep Kit II
Peqlab peqGOLD, Plasmid Purification, Plasmid Miniprep Kit II
  13-6945-00
 :  
undefined
Peqlab peqGOLD, Plasmid Purification, Plasmid Miniprep Kit II
  13-6945-00
 :  

 

  • Pk:
    5 Tests
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini plus
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    Plasmid Miniprep Kit II
  • Seq:
    01
  • Sample size:
    3 - 15 ml
  • Description:
    Plasmid Miniprep Kit II

 

 

Isolate up to 40 to 70 µg of high quality plasmid DNA from up to 15 ml bacterial cultures in 30 minutes or less. Simplified with VWR® peqGold Mini Column II technology into three quick steps: Bind, wash and elute.

  • Safe, no phenol/chloroform extractions
  • Versatile with both spin and vacuum formats available
  • High quality DNA is suitable for a wide range of downstream applications
 : Each kit contains peqGOLD DNA columns, 2 ml collection tubes, Solution I, Solution II, Solution III, HBC buffer, DNA wash buffer, RNase A and elution buffer.