Hydrochloric acid 6 M HCl (6N) volumetric solution, Fluka™
Catalog # X31087-2.5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:2,5 L
- Seq:5096
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (color):complying
- Appearance (form):complying
- APHA:Max. 20
- Assay:5.988 - 6.012 mol/l
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
:
:36.46 g/mol
UN:1789
:GHS07
:GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324
Density:1.072 g/cm³ (20 °C)
ADR:3 III