
Orthophosphoric acid ≥85%, Semiconductor grade (Honeywell 17861)
Catalog # HONC40266H-2.5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay:Min. 85 %
- Volatile acids (CH₃COOH):Max. 10 ppm
- Aluminium (Al):Max. 10 ppm
- Arsenic (As):Max. 0.5 ppm
- Calcium (Ca):Max. 20 ppm
- Cadmium (Cd):Max. 5 ppm
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 10 ppm
- Potassium (K):Max. 5 ppm
- Magnesium (Mg):Max. 10 ppm
- Manganese (Mn):Max. 0.5 ppm
- Sodium (Na):Max. 10 ppm
- Nickel (Ni):Max. 5 ppm
- Lead (Pb):Max. 5 ppm
- Antimony (Sb):Max. 2 ppm
- Zinc (Zn):Max. 10 ppm
- Chloride (Cl):Max. 1 ppm
- Nitrate (NO₃):Max. 5 ppm
- Sulfate (SO₄):Max. 30 ppm
- KMnO₄ reducing matter (as O):Max. 10 ppm
Specification Test Results
About this item
:
:98 g/mol
Boiling Pt:158 °C (1.013 hPa)
Melting Pt:21 °C
Density:1.71 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1805
ADR:8,III
:GHS05
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340