Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Hydrochloric acid fuming 37%, Semiconductor grade (Honeywell 17823)
  HONC40253H-2.5L
 :  
 
 
Hydrochloric acid fuming 37%, Semiconductor grade (Honeywell 17823)
  HONC40253H-2.5L
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Assay:
    37 - 38 %
  • Residue on ignition (SO₄):
    Max. 5 ppm
  • Ammonium (NH₄):
    Max. 2 ppm
  • Silver (Ag):
    Max. 0.02 ppm
  • Aluminium (Al):
    Max. 0.05 ppm
  • Arsenic (As):
    Max. 0.01 ppm
  • Gold (Au):
    Max. 0.02 ppm
  • Boron (B):
    Max. 0.02 ppm
  • Barium (Ba):
    Max. 0.1 ppm
  • Beryllium (Be):
    Max. 0.01 ppm
  • Bismuth (Bi):
    Max. 0.02 ppm
  • Calcium (Ca):
    Max. 0.2 ppm
  • Cadmium (Cd):
    Max. 0.01 ppm
  • Cobalt (Co):
    Max. 0.01 ppm
  • Chromium (Cr):
    Max. 0.01 ppm
  • Copper (Cu):
    Max. 0.01 ppm
  • Iron (Fe):
    Max. 0.2 ppm
  • Gallium (Ga):
    Max. 0.02 ppm
  • Indium (In):
    Max. 0.02 ppm
  • Potassium (K):
    Max. 0.1 ppm
  • Lithium (Li):
    Max. 0.02 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.01 ppm
  • Molybdenum (Mo):
    Max. 0.01 ppm
  • Sodium (Na):
    Max. 0.5 ppm
  • Nickel (Ni):
    Max. 0.01 ppm
  • Lead (Pb):
    Max. 0.02 ppm
  • Platinum (Pt):
    Max. 0.02 ppm
  • Antimony (Sb):
    Max. 0.01 ppm
  • Tin (Sn):
    Max. 0.02 ppm
  • Strontium (Sr):
    Max. 0.02 ppm
  • Titanium (Ti):
    Max. 0.01 ppm
  • Thallium (Tl):
    Max. 0.02 ppm
  • Vanadium (V):
    Max. 0.01 ppm
  • Zinc (Zn):
    Max. 0.05 ppm
  • Zirconium (Zr):
    Max. 0.01 ppm
  • Bromide (Br):
    Max. 50 ppm
  • Free chlorine (Cl):
    Max. 0.5 ppm
  • Phosphate (PO₄):
    Max. 0.5 ppm
  • Sulfate (SO₄):
    Max. 1 ppm
  • Sulfite (SO₃):
    Max. 1 ppm

 

 

 :
 :42 °C (1.013 hPa)
 :–35 °C
 :1.190 g/cm³ (20 °C)
 :Ambient
 :8,II
 :36.46 g/mol
 :1789
 :GHS07
 :GHS05
 :7647-01-0
 :HCl
 :MFCD00011324